missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-Dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4510441
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
368869-97-0
C12H9NO3S
247.27
MFCD01313485
NOUOFPSRFBTJCH-UHFFFAOYSA-N
2-2,3-dihydro-1-benzofuran-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dihydro-5-benzofuranyl, maybridge1_008369, 2-2,3-dihydrobenzofuran-5-yl-4-thiazolecarboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-1,3-thiazole-4-carboxylic acid
2742272
2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1
This item is not returnable. View return policy
368869-97-0
C12H9NO3S
247.27
MFCD01313485
NOUOFPSRFBTJCH-UHFFFAOYSA-N
2-2,3-dihydro-1-benzofuran-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dihydro-5-benzofuranyl, maybridge1_008369, 2-2,3-dihydrobenzofuran-5-yl-4-thiazolecarboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-1,3-thiazole-4-carboxylic acid
2742272
2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 247.27
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dihydro-1-benzofuran-5-yl)-1, 3-thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'