missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 274.13 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2, 3-Dichlorophenyl)-1, 3-thiazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'