missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3-Tetrafluoro-1-propanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-3830567
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
76-37-9
C3H4F4O
132.06
MFCD00004676
NBUKAOOFKZFCGD-UHFFFAOYSA-N
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
6441
2,2,3,3-tetrafluoropropan-1-ol
OCC(F)(F)C(F)F
This item is not returnable. View return policy
76-37-9
C3H4F4O
132.06
MFCD00004676
NBUKAOOFKZFCGD-UHFFFAOYSA-N
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
6441
2,2,3,3-tetrafluoropropan-1-ol
OCC(F)(F)C(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 1.4700g/mL
Boiling Point 109°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula F2CHCF2CH2OH
Refractive Index 1.3200 to 1.322
Quantity 25 g
Merck Index 15, 9351
Specific Gravity 1.47
Formula Weight 132.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'