missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%, Thermo Scientific Chemicals

Catalog Number 11480621
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
355-80-6
C5H4F8O
232.07
MFCD00039631
JUGSKHLZINSXPQ-UHFFFAOYSA-N
2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
9641
2,2,3,3,4,4,5,5-octafluoropentan-1-ol
OCC(F)(F)C(F)(F)C(F)(F)C(F)F
This item is not returnable. View return policy
355-80-6
C5H4F8O
232.07
MFCD00039631
JUGSKHLZINSXPQ-UHFFFAOYSA-N
2,2,3,3,4,4,5,5-octafluoro-1-pentanol, 1h,1h,5h-octafluoro-1-pentanol, 1-pentanol, 2,2,3,3,4,4,5,5-octafluoro, 1,1,5-trihydrooctafluoropentyl alcohol, 2,2,3,3,4,4,5,5-octafluoropentanol, 1,1,5-trihydrooctafluoropentan-1-ol, unii-d05k8ejx15, 1h,1h,5h-octafluoropentanol, 2,2,3,3,4,4,5,5-octafluoropentyl alcohol, alpha,alpha,omega-trihydroperfluoropentanol
9641
2,2,3,3,4,4,5,5-octafluoropentan-1-ol
OCC(F)(F)C(F)(F)C(F)(F)C(F)F

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

Solubility
Insoluble in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY
Melting Point <-50°C
Density 1.667
Boiling Point 141°C to 142°C
Flash Point 75°C (167°F)
Refractive Index 1.318
Quantity 500 g
Beilstein 1773494
Merck Index 14,6746
Solubility Information Insoluble in water.
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'