missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.
Solubility
Insoluble in water.
Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol is used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.
Solubility
Insoluble in water.
Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
Specifications
Melting Point | <-50°C |
Density | 1.667 |
Boiling Point | 141°C to 142°C |
Flash Point | 75°C (167°F) |
Refractive Index | 1.318 |
Quantity | 25 g |
Beilstein | 1773494 |
Merck Index | 14,6746 |
Solubility Information | Insoluble in water. |
Formula Weight | 232.07 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'