missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-iodobutane, 96%, Thermo Scientific Chemicals

Catalog Number p-7044035
16.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374-98-1
C4H2F7I
309.953
MFCD00013719
AFHPVXVEXINDFS-UHFFFAOYSA-N
heptafluorobutyl iodide, 2,2,3,3,4,4,4-heptafluoro-1-iodobutane, butane, 1,1,1,2,2,3,3-heptafluoro-4-iodo, 1h,1h-heptafluorobutyl iodide, 1h,1h-heptafluoro-1-iodobutane, acmc-20aoko, 1-iodo-1h,1h-perfluorobutane, 1h,1h-heptafluoro-1-iodobutan, 1-iodo-2,2,3,3,4,4,4-heptafluorobutane
67806
1,1,1,2,2,3,3-heptafluoro-4-iodobutane
C(C(C(C(F)(F)F)(F)F)(F)F)I
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
374-98-1
C4H2F7I
309.953
MFCD00013719
AFHPVXVEXINDFS-UHFFFAOYSA-N
heptafluorobutyl iodide, 2,2,3,3,4,4,4-heptafluoro-1-iodobutane, butane, 1,1,1,2,2,3,3-heptafluoro-4-iodo, 1h,1h-heptafluorobutyl iodide, 1h,1h-heptafluoro-1-iodobutane, acmc-20aoko, 1-iodo-1h,1h-perfluorobutane, 1h,1h-heptafluoro-1-iodobutan, 1-iodo-2,2,3,3,4,4,4-heptafluorobutane
67806
1,1,1,2,2,3,3-heptafluoro-4-iodobutane
C(C(C(C(F)(F)F)(F)F)(F)F)I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 2.054
Boiling Point 91°C to 92°C
Flash Point None
Linear Formula CF3CF2CF2CH2I
Refractive Index 1.3625
Quantity 1 g
Beilstein 1761927
Sensitivity Light sensitive
Formula Weight 309.95
Percent Purity 96%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'