missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals
54.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Description
It is employed as a halogenated solvent in electrospray liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a halogenated solvent in electrospray liquid chromatography.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
It is employed as a halogenated solvent in electrospray liquid chromatography.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Specifications
Density | 1.596 |
Boiling Point | 95°C to 97°C |
Flash Point | 25°C (77°F) |
Linear Formula | CF3CF2CF2CH2OH |
Refractive Index | 1.3 |
Quantity | 25 g |
UN Number | UN1987 |
Beilstein | 1761907 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 200.06 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'