missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine, 99.5%, Thermo Scientific Chemicals

Catalog Number p-3737646
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
753-90-2
C2H4F3N
99.06
MFCD00008132
KIPSRYDSZQRPEA-UHFFFAOYSA-N
2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine
9773
2,2,2-trifluoroethanamine
C(C(F)(F)F)N
This item is not returnable. View return policy
753-90-2
C2H4F3N
99.06
MFCD00008132
KIPSRYDSZQRPEA-UHFFFAOYSA-N
2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine
9773
2,2,2-trifluoroethanamine
C(C(F)(F)F)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.4
CAS Max % 100.0
Color Colorless
Density 1.2400g/mL
Boiling Point 37°C to 38°C
Flash Point -16°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula CF3CH2NH2
Refractive Index 1.3000 to 1.302
Quantity 1 g
Specific Gravity 1.24
Formula Weight 99.06
Percent Purity 99.50%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'