missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2,2-Trifluoroethyl methanesulfonate, 97%

Brand:  Alfa Aesar™ H64729.14

Code : NEW

Additional Details : CAS Number : 25236-64-0 Weight : 0.02500kg Transport : UN number : 3265 Chem class : 1a Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15412039

  • £125.08 / 25g


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

2,2,2-Trifluoroethyl methanesulfonate
H3CSO2OCH2CF3
MFCD00025058
1863049
25236-64-0
178.13
UN3265
2,2,2-Trifluoromethyl mesylate

Specifications

2,2,2-Trifluoroethyl methanesulfonate
Moisture Sensitive
25g
1.3670
97%
178.13
UN3265
2,2,2-Trifluoromethyl mesylate
1.513g/mL
Colorless
25236-64-0
Liquid
H3CSO2OCH2CF3
MFCD00025058
1863049
Videos
SDS