missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-204458
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
352-87-4
C6H7F3O2
168.12
MFCD00013576,MFCD00274739
QTKPMCIBUROOGY-UHFFFAOYSA-N
2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
9608
2,2,2-trifluoroethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
352-87-4
C6H7F3O2
168.12
MFCD00013576,MFCD00274739
QTKPMCIBUROOGY-UHFFFAOYSA-N
2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
9608
2,2,2-trifluoroethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color White
Density 1.1800g/mL
Boiling Point 59.0°C (100.0 mmHg)
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CF3
Refractive Index 1.3600 to 1.3620
Quantity 25 g
Specific Gravity 1.18
Stabilizer 100ppm MEHQ
Formula Weight 168.12
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'