missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol-d{3}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number 11386079
268.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 x 1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
77253-67-9
C2H3F3O
103.06
MFCD00037682
RHQDFWAXVIIEBN-IDPMSXFZSA-N
2,2,2-trifluoroethanol-d3, 1,1-2h2-2,2,2-trifluoroetane-1-2h ol, ethan-1,1-d2-ol-d, 2,2,2-trifluoro, trifluoro 2 h? ethan 2 h ol, 2,2,2-trifluoroethanol-d3 >98 atom % d, 2,2,2-trifluoroethanol-d3, ≥99.5 atom % d, 2,2,2-trifluoroethanol-d3, 99 atom % d cp
2724851
[2H]OC([2H])([2H])C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
77253-67-9
C2H3F3O
103.06
MFCD00037682
RHQDFWAXVIIEBN-IDPMSXFZSA-N
2,2,2-trifluoroethanol-d3, 1,1-2h2-2,2,2-trifluoroetane-1-2h ol, ethan-1,1-d2-ol-d, 2,2,2-trifluoro, trifluoro 2 h? ethan 2 h ol, 2,2,2-trifluoroethanol-d3 >98 atom % d, 2,2,2-trifluoroethanol-d3, ≥99.5 atom % d, 2,2,2-trifluoroethanol-d3, 99 atom % d cp
2724851
[2H]OC([2H])([2H])C(F)(F)F

NMR Solvent

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.45 g/mL
Packaging Prescored Ampule
Sensitivity Hygroscopic
Quantity 5 x 1 g
Melting Point -44°C
Boiling Point 77°C to 80°C
UN Number UN1992
IUPAC Name 1,1-dideuterio-1-deuteriooxy-2,2,2-trifluoroethane
Formula Weight 103.06
Percent Purity 99%
Chemical Name or Material 2,2,2-Trifluoroethanol-d3, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'