missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number 10708542
Click to view available options
Quantity:
1 g
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1648-99-3
C2H2ClF3O2S
182.55
MFCD00007458
CXCHEKCRJQRVNG-UHFFFAOYSA-N
tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
74242
2,2,2-trifluoroethanesulfonyl chloride
C(C(F)(F)F)S(=O)(=O)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1648-99-3
C2H2ClF3O2S
182.55
MFCD00007458
CXCHEKCRJQRVNG-UHFFFAOYSA-N
tresyl chloride, 2,2,2-trifluoroethane-1-sulfonyl chloride, ethanesulfonyl chloride, 2,2,2-trifluoro, 2,2,2-trifluoroethanesulphonyl chloride, 2,2,2-trifluoroethyl sulphonyl chloride, 2,2,2-trifluoroethylsulfonyl chloride, chloro 2,2,2-trifluoroethyl sulfone, 2,2,2-trifluoroethanesulfonylchloride, ethanesulfonylchloride, 2,2,2-trifluoro, pubchem2553
74242
2,2,2-trifluoroethanesulfonyl chloride
C(C(F)(F)F)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.6500g/mL
Boiling Point 64°C to 67°C (45.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula CF3CH2SO2Cl
Refractive Index 1.3870 to 1.389
Quantity 2.5 g
Specific Gravity 1.65
Formula Weight 182.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'