missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog Number 10785213
Click to view available options
Quantity:
25 g
250 g
2.5 kg
This item is not returnable. View return policy
354-38-1
C2H2F3NO
113.04
MFCD00008008
NRKYWOKHZRQRJR-UHFFFAOYSA-N
trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
67717
2,2,2-trifluoroacetamide
NC(=O)C(F)(F)F
This item is not returnable. View return policy
354-38-1
C2H2F3NO
113.04
MFCD00008008
NRKYWOKHZRQRJR-UHFFFAOYSA-N
trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
67717
2,2,2-trifluoroacetamide
NC(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 70°C
Color White to Yellow or Beige
Boiling Point 162°C to 164°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CF3CONH2
Quantity 25 g
Beilstein 02, II, 186
Solubility Information Solubility in water: 460g/l (20°C). Other solubilities: soluble in alcohol,ether,partially soluble in chloroform
Formula Weight 113.04
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 2, 2-Trifluoroacetamide
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,2-Trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'