missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-203076
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
115-20-8
C2H3Cl3O
149.4
MFCD00004677
KPWDGTGXUYRARH-UHFFFAOYSA-N
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
8259
CHEBI:28094
2,2,2-trichloroethanol
C(C(Cl)(Cl)Cl)O
115-20-8
C2H3Cl3O
149.4
MFCD00004677
KPWDGTGXUYRARH-UHFFFAOYSA-N
trichloroethanol, trichlorethanol, trichloroethyl alcohol, ethanol, 2,2,2-trichloro, 2,2,2-trichloro-1-ethanol, hydroxymethyl trichloromethane, 2,2,2-trichloroethyl alcohol, 2,2,2-trichloroethan-1-ol, unii-aw835aj62n, beta-trichloroethanol
8259
CHEBI:28094
2,2,2-trichloroethanol
C(C(Cl)(Cl)Cl)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 17.8°C
Color Colorless to Yellow
Density 1.5500g/mL
Boiling Point 151.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CCl3CH2OH
Refractive Index 1.4885 to 1.4905
Quantity 100 g
Beilstein 01, 338
Fieser 03,295; 04,521; 06,605
Merck Index 15, 9803
Specific Gravity 1.55
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether and carbon,tetrachloride
Formula Weight 149.4
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trichloroethanol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trichloroethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'