missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroacetamide, 98+%, Thermo Scientific Chemicals

Catalog Number p-3779696
10.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
594-65-0
C2H2Cl3NO
162.39
MFCD00008009
UPQQXPKAYZYUKO-UHFFFAOYSA-N
trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
61144
2,2,2-trichloroacetamide
NC(=O)C(Cl)(Cl)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
594-65-0
C2H2Cl3NO
162.39
MFCD00008009
UPQQXPKAYZYUKO-UHFFFAOYSA-N
trichloroacetamide, acetamide, 2,2,2-trichloro, amid kyseliny trichloroctove, acetamide, alpha-trichloro, unii-i8bl3305ro, amid kyseliny trichloroctove czech, 2,2,2-trichloro-acetamide, acetamide, .alpha.-trichloro, .alpha.,.alpha.,.alpha.-trichloroacetamide, trichloro-acetamide
61144
2,2,2-trichloroacetamide
NC(=O)C(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5, 98%
Melting Point 139°C to 143°C
Color White
Boiling Point 238°C to 240°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula CCl3CONH2
Quantity 25 g
Beilstein 02, 211
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in ethanol,ethylether
Formula Weight 162.4
Percent Purity 98+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2, 2, 2-Trichloroacetamide
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,2-Trichloroacetamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'