missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloro-1-ethoxyethanol, 85%, Tech., ACROS Organics™

Brand:  Acros Organics 346530250

Code : 29

Additional Details : CAS Number : 515-83-3 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10347461

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 2, 2-Trichloro-1-ethoxyethanol
80.0
Technical Grade
C4H7Cl3O2
MFCD00013668
515-83-3
100.0
85%
193.46

Specifications

2, 2, 2-Trichloro-1-ethoxyethanol
515-83-3
100.0
Crystalline Powder, Crystals, Needles or Low Melting Mass
85%
Authentic
MFCD00013668
Chloral alcoholate
Glass bottle
116.0°C
White to light yellow
25g
80.0
Technical Grade
85%
C4H7Cl3O2
193.46
15, 2069
80% min.
3% max. (K.F.)
47.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,2,2-Trichloro-1-ethoxyethanol, tech.

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS