missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3737602
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
75-80-9
C2H3Br3O
282.76
MFCD00004671
YFDSDPIBEUFTMI-UHFFFAOYSA-N
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
6400
2,2,2-tribromoethanol
C(C(Br)(Br)Br)O
This item is not returnable. View return policy
75-80-9
C2H3Br3O
282.76
MFCD00004671
YFDSDPIBEUFTMI-UHFFFAOYSA-N
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
6400
2,2,2-tribromoethanol
C(C(Br)(Br)Br)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 73°C to 79°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br3CCH2OH
Quantity 10 g
Beilstein 01, II, 338
Fieser 11,111
Merck Index 12, 9743
Solubility Information Solubility in water: 25g/L @ 40°C
Formula Weight 282.76
Percent Purity 99%
Chemical Name or Material 2, 2, 2-Tribromoethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,2-Tribromoethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'