missing translation for 'onlineSavingsMsg'
Learn More

2-[(2,2-Dimethylpropanoyl)amino]nicotinic acid, 97%, Thermo Scientific™

Catalog Number 10623413
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
125867-25-6
C11H14N2O3
222.244
MFCD03659710
KXLOGMIKCNUSNG-UHFFFAOYSA-N
2-pivalamidonicotinic acid, 2-2,2-dimethyl-propionylamino-nicotinic acid, 2-2,2,2-trimethylacetamido nicotinic acid, 2-2,2-dimethylpropanoyl amino nicotinic acid, 2-2,2-dimethylpropanamido pyridine-3-carboxylic acid, 2-pivaloylamino nicotinic acid, 2-2,2-dimethylpropanoylamino pyridine-3-carboxylic acid, maybridge3_004279, acmc-20aorg, pubchem16551
2779758
2-(2,2-dimethylpropanoylamino)pyridine-3-carboxylic acid
CC(C)(C)C(=O)NC1=C(C=CC=N1)C(=O)O
This item is not returnable. View return policy
125867-25-6
C11H14N2O3
222.244
MFCD03659710
KXLOGMIKCNUSNG-UHFFFAOYSA-N
2-pivalamidonicotinic acid, 2-2,2-dimethyl-propionylamino-nicotinic acid, 2-2,2,2-trimethylacetamido nicotinic acid, 2-2,2-dimethylpropanoyl amino nicotinic acid, 2-2,2-dimethylpropanamido pyridine-3-carboxylic acid, 2-pivaloylamino nicotinic acid, 2-2,2-dimethylpropanoylamino pyridine-3-carboxylic acid, maybridge3_004279, acmc-20aorg, pubchem16551
2779758
2-(2,2-dimethylpropanoylamino)pyridine-3-carboxylic acid
CC(C)(C)C(=O)NC1=C(C=CC=N1)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-[(2, 2-Dimethylpropanoyl)amino]nicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'