missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2-chloroethoxy)ethyl]-1H-isoindole-1,3(2H)-dione, 97%, Maybridge

Brand:  Maybridge KM10098EA

Code : 31

Additional Details : CAS Number : 58290-51-0 Weight : 0.06000kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10384192

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
97.0
97%
253.68
58290-51-0
97%
C12H12ClNO3
MFCD00196075

Specifications

2-[2-(2-Chloroethoxy)ethyl]-1H-isoindole-1, 3(2H)-dione
58290-51-0
Amber Glass Bottle
97%
253.68
10g
97.0
97%
C12H12ClNO3
MFCD00196075
Videos
SDS