missing translation for 'onlineSavingsMsg'
Learn More

2(1H)-Quinolinone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15416327
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
91301-03-0
C10H7NO2
173.17
MFCD00563443
VWHKEYXRRNSJTN-UHFFFAOYSA-N
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril
589334
2-oxo-1H-quinoline-3-carbaldehyde
O=CC1=CC2=CC=CC=C2NC1=O
This item is not returnable. View return policy
91301-03-0
C10H7NO2
173.17
MFCD00563443
VWHKEYXRRNSJTN-UHFFFAOYSA-N
2-hydroxyquinoline-3-carbaldehyde, 2-oxo-1,2-dihydro-quinoline-3-carbaldehyde, 2-oxo-1,2-dihydroquinoline-3-carbaldehyde, 3-formyl-2quinolone, 1,2-dihydro-2-oxoquinoline-3-carboxaldehyde, 1,2-dihydro-2-oxoquinoline-3-carbaldehyde, 2-hydroxy-3-quinolinecarboxaldehyde, 2-oxo-1,2-dihydro-3-quinolinecarbaldehyde, 2-oxohydroquinoline-3-carbaldehyde, 3-formylcarbostyril
589334
2-oxo-1H-quinoline-3-carbaldehyde
O=CC1=CC2=CC=CC=C2NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >250°C
Color White to Brown
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 173.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2(1H)-Quinolinone-3-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'