Learn More
Thermo Scientific Maybridge 2-(1H-Pyrrol-1-yl)benzoic acid, 5g, 97%
Brand: Thermo Scientific Maybridge CC34301DE
2-(1H-Pyrrol-1-yl)benzoic acid, 5g, 97%
Code : 31
Additional Details : CAS Number : 10333-68-3 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CC34301DE
2-(1H-Pyrrol-1-yl)benzoic acid, 5g, 97%
Code : 31
Additional Details : CAS Number : 10333-68-3 Weight : 0.05500kg