missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)benzylamine, 97%, ACROS Organics™

Brand:  Acros Organics 444090050

Code : 29

Additional Details : CAS Number : 25373-55-1 Weight : 0.05500kg Transport : UN number : 3259 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 11936281

  • £585.70 / 5g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

2-(1H-Imidazol-1-yl)benzylamine
97%
173.22
≥96.0%
C10H11N3
1-(α-Amino-o-tolyl)imidazole

Specifications

2-(1H-Imidazol-1-yl)benzylamine
Authentic
≥96.0%
C10H11N3
Pale yellow
5g
25373-55-1
Crystalline Powder
97%
173.22
50° to 54°C
1-(α-Amino-o-tolyl)imidazole
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(1H-Imidazol-1-yl)benzylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS