missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Imidazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog Number p-3484841
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
26286-54-4
C9H9N3
159.192
MFCD03274531
HVECTIQVQPUSEX-UHFFFAOYSA-N
2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
320166
2-imidazol-1-ylaniline
C1=CC=C(C(=C1)N)N2C=CN=C2
This item is not returnable. View return policy
26286-54-4
C9H9N3
159.192
MFCD03274531
HVECTIQVQPUSEX-UHFFFAOYSA-N
2-1h-imidazol-1-yl aniline, 2-1-imidazolyl aniline, 2-imidazol-1-yl-phenylamine, 2-imidazol-1-yl aniline, benzenamine,2-1h-imidazol-1-yl, 2-imidazolylphenylamine, 2-1h-imidazol-1-yl benzenamine, pubchem10103, n-o-aminophenyl imidazole, 2-imidazol-1-ylphenylamine
320166
2-imidazol-1-ylaniline
C1=CC=C(C(=C1)N)N2C=CN=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 2-(1H-Imidazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'