missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-1,2,4-triazol-1-ylmethyl)aniline, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Specifica
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 174.21 |
Percent Purity | 97% |
Chemical Name or Material | 2-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'