missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals
22.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.
Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.
Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | 257°C to 261°C |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 3716843 |
Merck Index | 14,6368 |
Solubility Information | Soluble in water 170g/L,alcohol,DMSO,ethanol. |
Formula Weight | 246.74 |
Percent Purity | 99% |
Chemical Name or Material | 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'