missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Oxazol-5-yl)aniline, ≥97%, Thermo Scientific™

Catalog Number 10729413
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
774238-36-7
C9H8N2O
160.176
MFCD07772822
UZPQWOKKMYUKNI-UHFFFAOYSA-N
7162068
2-(1,3-oxazol-5-yl)aniline
C1=CC=C(C(=C1)C2=CN=CO2)N
This item is not returnable. View return policy
774238-36-7
C9H8N2O
160.176
MFCD07772822
UZPQWOKKMYUKNI-UHFFFAOYSA-N
7162068
2-(1,3-oxazol-5-yl)aniline
C1=CC=C(C(=C1)C2=CN=CO2)N
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-1,3-oxazol-5-yl aniline, 2-5-oxazolyl aniline, 2-oxazol-5-yl aniline, benzenamine, 2-5-oxazolyl, 2-oxazol-5-yl phenylamine, benzenamine,2-5-oxazolyl, 5-2-aminophenyl-1,3-oxazole, 2-1,3-oxazol-5-yl phenylamine
Formula Weight 160.18
Percent Purity 90%
Chemical Name or Material 2-(1, 3-Oxazol-5-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'