missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazol-5-yl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-2696541
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
337508-62-0
C7H3N3S2
193.242
MFCD02681895
HHDWFDQYUFKNCO-UHFFFAOYSA-N
2776292
5-isothiocyanato-2,1,3-benzothiadiazole
C1=CC2=NSN=C2C=C1N=C=S
This item is not returnable. View return policy
337508-62-0
C7H3N3S2
193.242
MFCD02681895
HHDWFDQYUFKNCO-UHFFFAOYSA-N
2776292
5-isothiocyanato-2,1,3-benzothiadiazole
C1=CC2=NSN=C2C=C1N=C=S
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 2,1,3-benzothiadiazol-5-yl isothiocyanate, 2,1,3-benzothiadiazol-5-ylisothiocyanate, 2,1,3-benzothiadiazole, 5-isothiocyanato, 2,1,3-benzothiadiazole,5-isothiocyanato, benzo c 1,2,5-thiadiazol-5-isothiocyanate, 5-isothiocyanatobenzo c 1.2.5 thiadiazole
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazol-5-yl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'