missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzothiadiazol-5-yl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | 2,1,3-benzothiadiazol-5-yl isothiocyanate, 2,1,3-benzothiadiazol-5-ylisothiocyanate, 2,1,3-benzothiadiazole, 5-isothiocyanato, 2,1,3-benzothiadiazole,5-isothiocyanato, benzo c 1,2,5-thiadiazol-5-isothiocyanate, 5-isothiocyanatobenzo c 1.2.5 thiadiazole |
Formula Weight | 193.25 |
Percent Purity | 97% |
Chemical Name or Material | 2, 1, 3-Benzothiadiazol-5-yl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'