missing translation for 'onlineSavingsMsg'
Learn More

2-(1,1-Difluoroethyl)pyrazine, 97%, Thermo Scientific™

Catalog Number p-4812635
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
111781-49-8
C6H6F2N2
144.12
OXKFWGSACYGCSY-UHFFFAOYSA-N
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl
19763564
2-(1,1-difluoroethyl)pyrazine
CC(C1=NC=CN=C1)(F)F
This item is not returnable. View return policy
111781-49-8
C6H6F2N2
144.12
OXKFWGSACYGCSY-UHFFFAOYSA-N
2-1,1-difluoroethyl pyrazine, 3-1,1-difluoroethyl pyrazine, pyrazine, 2-1,1-difluoroethyl
19763564
2-(1,1-difluoroethyl)pyrazine
CC(C1=NC=CN=C1)(F)F
Boiling Point 150.0°C to 152.0°C
Flash Point 53°C
Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 144.12
Percent Purity 97%
Chemical Name or Material 2-(1, 1-Difluoroethyl)pyrazine
compliance-icons compliance-icons
Product Identifier
  • 2-(1,1-Difluoroethyl)pyrazine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'