missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-Verbenone, 94%, ACROS Organics™

Brand:  Acros Organics 203630100

Code : 29

Additional Details : CAS Number : 1196-01-6 Weight : 0.01000kg

Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10522611

  • £58.10 / 10g


EU Stock 7
Add to basket

Specifications

Chemical Identifiers

(1S)-(-)-Verbenone
1196-01-6
94%
150.22
(1S)-(-)-Verbenone
94%
≥92%
C10H14O
MFCD00065445

Specifications

(1S)-(-)-Verbenone
1196-01-6
Glass bottle
Liquid
94%
-150° to -250° (20°C, 589nm) (C=4, CHCl3) ,reported only for information
MFCD00065445
07, 161
(1S)-(-)-Verbenone
Clear light yellow to yellow
94%
Authentic
1.4950 to 1.4970 (20°C, 589nm)
≥92%
C10H14O
150.22
0.96 to 0.98 (25°C)
0.9700g/mL
− 220.00 (20.00°C c=3−4,CHCL3)
10g
Videos
SDS