missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | -61.0°C |
Color | Colorless to Yellow |
Density | 0.8700g/mL |
Boiling Point | 165.0°C to 167.0°C |
Flash Point | 32°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- (1S)-(-)-ß-Pinene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H304-May be fatal if swallowed and enters airways.
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'