missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-β-Pinene, 98%, Thermo Scientific Chemicals

Catalog Number 15179183
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
18172-67-3
C10H16
136.24
MFCD00001345
WTARULDDTDQWMU-IUCAKERBSA-N
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
440967
CHEBI:28359
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
CC1(C2CCC(=C)C1C2)C
This item is not returnable. View return policy
18172-67-3
C10H16
136.24
MFCD00001345
WTARULDDTDQWMU-IUCAKERBSA-N
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
440967
CHEBI:28359
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
CC1(C2CCC(=C)C1C2)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -61.0°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 165.0°C to 167.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Metal drum
Refractive Index 1.4770 to 1.4810
Quantity 5 L
UN Number 2319
Beilstein 05,154
Merck Index 14,7446
Specific Gravity 0.87
Specific Rotation Condition − 19.50 (25.00°C c=neat)
Specific Rotation -19.5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 136.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (1S)-(-)-β-Pinene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (1S)-(-)-ß-Pinene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'