missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1S)-(+)-Menthyl glyoxylate monohydrate, 98%

Catalog Number p-7058519
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
C12H22O4
MFCD04037548
Glyoxylic acid (1S)-menthyl ester hydrate
This item is not returnable. View return policy
C12H22O4
MFCD04037548
Glyoxylic acid (1S)-menthyl ester hydrate
Melting Point 76°C to 80°C
Density 1.182
Boiling Point 95°C to 96°C (0.3 mmHg)
Quantity 5 g
UN Number UN3077
Beilstein 4905777
Sensitivity Air sensitive
Optical Rotation +74° (c=1 in Methanol)
Formula Weight 230.31 (212.29 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1S)-(+)-Menthyl glyoxylate monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'