missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Menthyl chloroformate, 97%, ACROS Organics™

Brand:  Acros Organics 314510250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 7635-54-3 Weight : 0.07550kg Transport : UN number : 3277 Chem class : 1a Pack group : II

Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10648634

  • £115.90 / 25mL


Estimated Shipment
11-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

(1S)-(+)-Menthyl chloroformate
7635-54-3
100.0
97%
218.73
97%
96.0
≥96.0%
C11H19ClO2
MFCD00134483

Specifications

(1S)-(+)-Menthyl chloroformate
Clear colorless to brown
97%
96.0
1.02g/mL
≥96.0%
C11H19ClO2
218.73
1.4570 to 1.4590 (20°C, 589nm)
+78° to +88° (20°C, 589nm) (c=1, CHCl3)
+83° (20°C c=1,CHCl3)
25mL
7635-54-3
100.0
Liquid
97%
Authentic
MFCD00134483
Glass bottle
Documentation

Technical Documents