missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%, Thermo Scientific™

£14.90 - £146.00

Identificadores de productos químicos

CAS 3144-16-9
Molecular Formula C10H15O4S
Molecular Weight (g/mol) 231.29
MDL Number MFCD00064157,MFCD00074827
InChI Key MIOPJNTWMNEORI-XVKPBYJWSA-M
Synonym 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
PubChem CID 65617
ChEBI CHEBI:55403
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
Ver más especificaciones

Productos 3
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
11453247
SDS Ver documentos
Thermo Scientific Alfa Aesar
A16179.14
25g
£14.90
25g
Ver productos alternativos
Ahorre hasta 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
11463247
SDS Ver documentos
Thermo Scientific Alfa Aesar
A16179.22
100g
£43.80
100g
Ver productos alternativos
Ahorre hasta 
»
In stock, ready to ship: 7
Please to purchase this item. Need a web account? Register with us today!
11473247
SDS Ver documentos
Thermo Scientific Alfa Aesar
A16179.36
500g
£146.00
500g
Ver productos alternativos
Ahorre hasta 
»
In stock, ready to ship: 7
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Applications
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

Solubility
Exhibit moderate solubility in HCCl.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.
Especificaciones

Identificadores de productos químicos

3144-16-9
231.29
MIOPJNTWMNEORI-XVKPBYJWSA-M
65617
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
C10H15O4S
MFCD00064157,MFCD00074827
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
CHEBI:55403
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Especificaciones

(1S)-(+)-Camphor-10-sulfonic acid
Hygroscopic
25g
(dry wt.), water <2%
MFCD00064157,MFCD00074827
2809675
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
MIOPJNTWMNEORI-XVKPBYJWSA-M
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
65617
232.3
+20° (c=2 in Water)
196°C to 200°C (decomposition)
3144-16-9
C10H15O4S
UN3261
14,1734
Exhibit moderate solubility in HCCl.
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
231.29
CHEBI:55403
≥98%
SDS
Product Certifications

RUO – Research Use Only