missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Camphor-10-sulfonic acid, 98+%(dry wt.), water <2%, Thermo Scientific Chemicals

Catalog Number 11463247
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3144-16-9
C10H15O4S
231.29
MFCD00064157,MFCD00074827
MIOPJNTWMNEORI-XVKPBYJWSA-M
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
65617
CHEBI:55403
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3144-16-9
C10H15O4S
231.29
MFCD00064157,MFCD00074827
MIOPJNTWMNEORI-XVKPBYJWSA-M
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
65617
CHEBI:55403
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S)-(+)-Camphor-10-sulfonic acid is used as stabilizer. (+)-(1S)-camphor-10-sulfonic acid being a very effective resolving agent. . Chiral polyaniline(PANI) nanofibers were synthesized via facilely potentiostatic electropolymerization method without template in the presence of(1S)-(+)camphor-10-sulfonic acid(D-CSA) or(1R)-(-)camphor-10-sulfonic acid(L-CSA) as the dopant. Synthesis of QUATs derived from (1S)-(+)camphor-10-sulfonic acid.

Solubility
Exhibit moderate solubility in HCCl.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases. Keep under dry inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 196°C to 200°C (decomposition)
Quantity 100 g
UN Number UN3261
Beilstein 2809675
Sensitivity Hygroscopic
Merck Index 14,1734
Solubility Information Exhibit moderate solubility in HCCl.
Optical Rotation +20° (c=2 in Water)
Formula Weight 232.3
Percent Purity ≥98%
Assay (dry wt.), water <2%
Chemical Name or Material (1S)-(+)-Camphor-10-sulfonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'