missing translation for 'onlineSavingsMsg'
Learn More

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, Thermo Scientific™

Catalog Number p-4510674
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
134003-03-5
UIVLZOWDXYXITH-UHNVWZDZSA-N
1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
2734523
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
C1CC2CC1C(=O)N2
This item is not returnable. View return policy
134003-03-5
UIVLZOWDXYXITH-UHNVWZDZSA-N
1s,4r-2-azabicyclo 2.2.1 heptan-3-one, 2-azabicyclo 2.2.1 heptan-3-one, 1s,4r, 1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one, 1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
2734523
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
C1CC2CC1C(=O)N2
Melting Point 76.2°C
Color White to Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H9NO
Quantity 250 mg
Molecular Weight (g/mol) 111.14
Formula Weight 111.14
Percent Purity 95%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'