missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-69546
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
161660-94-2
C11H18NO4
228.27
MFCD01320858,MFCD01320857
RNJQBGXOSAQQDG-SFYZADRCSA-M
1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
1512526
CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O
This item is not returnable. View return policy
161660-94-2
C11H18NO4
228.27
MFCD01320858,MFCD01320857
RNJQBGXOSAQQDG-SFYZADRCSA-M
1r,3s-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, --1r,3s-n-boc-3-aminocyclopentanecarboxylic acid, cis-3-boc-amino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3s-boc-3-aminocyclopentane-1-carboxylic acid, cis-3-boc-amino cyclopentane-1-carboxylic acid, --1r,3s-n-boc-3-aminocyclopentane carboxylic acid
1512526
CC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 96.1°C
CAS Min % 94.0
CAS Max % 100.0
Color White
Assay Percent Range 94% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
Formula Weight 229.27
Percent Purity 95%,98% ee
Quantity 250 mg
Physical Form Powder
Chemical Name or Material (1S, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'