missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, ACROS Organics™

Brand:  Acros Organics 434091000

Code : 29

Additional Details : CAS Number : 1091606-67-5 Weight : 0.05010kg


Quantity 100mg
Packaging Glass bottle

 View more versions of this product

Product Code. 11985931

  • £77.70 / 100mg


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

(1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
96.0
97%
381.45
1091606-67-5
100.0
C26H24NP

Specifications

(1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
87.0° to 93.0°C
1091606-67-5
100.0
96.0% min.
Crystalline Powder
97%
381.45
White to yellow
100mg
96.0
Authentic
Glass bottle
97%
C26H24NP
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS