missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-(-)-cis-1-Amino-2-indanol, 99%, ACROS Organics™

Brand:  Acros Organics 315650010

Code : 29

Additional Details : CAS Number : 126456-43-7 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10452461

  • £26.90 / 1g


Estimated Shipment
21-06-2018
Add to basket

Specifications

Chemical Identifiers

(1S, 2R)-(-)-cis-1-Amino-2-indanol
126456-43-7
100.0
99%
149.19
99%
98.5
≥98.5%
C9H11NO
MFCD00216655

Specifications

(1S, 2R)-(-)-cis-1-Amino-2-indanol
126456-43-7
100.0
Glass bottle
Powder
99%
C9H11NO
149.19
White to light beige
99%
98.5
Authentic
-42.5° to -47.5° (20°C, 589nm) (c=1, CH3OH)
≥98.5%
− 62.00 (20.00°C c=0.5,CHCl3)
117° to 121°C
MFCD00216655
1g
Videos
SDS