missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol hydrochloride ≥97%

Brand:  Alfa Aesar™ H34178.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15468815

  • £336.00 / 25g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol hydrochloride
241.76
MFCD09749874
β-Methyl-α-phenyl-1-pyrrolidineethanol hydrochloride; N-Pyrrolidinyl-d-norephedrine hydrochloride

Specifications

(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)-1-propanol hydrochloride
White to cream
25g
≥97%
MFCD09749874
Hygroscopic
222° to 224°C
Powder
241.76
β-Methyl-α-phenyl-1-pyrrolidineethanol hydrochloride; N-Pyrrolidinyl-d-norephedrine hydrochloride
Videos
SDS