missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Menthyl chloroformate, 90+%, Thermo Scientific™

Catalog Number p-7058376
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14602-86-9
C11H19ClO2
218.72
MFCD00009694,MFCD00190089
KIUPCUCGVCGPPA-UHFFFAOYNA-N
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
7014897
CC(C)C1CCC(C)CC1OC(Cl)=O
This item is not returnable. View return policy
14602-86-9
C11H19ClO2
218.72
MFCD00009694,MFCD00190089
KIUPCUCGVCGPPA-UHFFFAOYNA-N
1r---menthyl chloroformate, --menthyl chloroformate, 1r,2s,5r-2-isopropyl-5-methylcyclohexyl carbonochloridate, --menthylchloroformate, pubchem8110, ?-menthyl chloroformate, --chloroformyloxy-p-menthane, 1r,2s,5r-2-isopropyl-5-methylcyclohexylcarbonochloridate, 1r,2s,5r-5-methyl-2-propan-2-yl cyclohexyl chloroformate, 1r---menthyl chloroformate, optical purity ee glc
7014897
CC(C)C1CCC(C)CC1OC(Cl)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.025
Boiling Point 108°C to 109°C (11 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.458
Quantity 5 g
UN Number UN3277
Beilstein 2414686
Sensitivity Moisture sensitive
Optical Rotation −82° (c=1 in Chloroform)
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
Formula Weight 218.73
Percent Purity 95%
Chemical Name or Material (1R)-(-)-Menthyl chloroformate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'