missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-(-)-Camphor-10-sulfonic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.
Solubility
Soluble in water, and a wide variety of organic substances.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids and bases.
(1R)-(-)-Camphor-10-sulfonic acid, is used as a pharamaceutical intermediate and also used as a chiral derivative of Camp. It can also be used as resolving agents for chiral amines and other cations.
Solubility
Soluble in water, and a wide variety of organic substances.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong acids and bases.
Specifications
Melting Point | ∼200°C |
Quantity | 25 g |
UN Number | UN3261 |
Beilstein | 2809676 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water,and a wide variety of organic substances. |
Optical Rotation | −21° (c=10 in Water) |
IUPAC Name | [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid |
Formula Weight | 232.3 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'