missing translation for 'onlineSavingsMsg'
Learn More

(1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3770786
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
60410-16-4
C7H10O3
142.15
MFCD00078933
IJDYOKVVRXZCFD-RQJHMYQMSA-N
1s,4r-cis-4-acetoxy-2-cyclopenten-1-ol, 1r,4s-4-hydroxycyclopent-2-en-1-yl acetate, 1r,4s-4-hydroxycyclopent-2-enyl acetate, 1r,3s-+-cis-4-cyclopentene-1,3-diol 1-acetate, acetic acid 1r,4s-4-hydroxy-cyclopent-2-enyl ester, 1r,3s-+-4-cyclopentene-1,3-diol 1-acetate, 1s,4r-4-acetoxy-2-cyclopenten-1-ol, 1r,3s-4-cyclopentene-1,3-diol 1-acetate, 1r,4s-cis-4-hydroxy-2-cyclopentenyl acetate, 5beta-acetoxycyclopentene-3beta-ol
10154103
[(1R,4S)-4-hydroxycyclopent-2-en-1-yl] acetate
CC(=O)OC1CC(C=C1)O
This item is not returnable. View return policy
60410-16-4
C7H10O3
142.15
MFCD00078933
IJDYOKVVRXZCFD-RQJHMYQMSA-N
1s,4r-cis-4-acetoxy-2-cyclopenten-1-ol, 1r,4s-4-hydroxycyclopent-2-en-1-yl acetate, 1r,4s-4-hydroxycyclopent-2-enyl acetate, 1r,3s-+-cis-4-cyclopentene-1,3-diol 1-acetate, acetic acid 1r,4s-4-hydroxy-cyclopent-2-enyl ester, 1r,3s-+-4-cyclopentene-1,3-diol 1-acetate, 1s,4r-4-acetoxy-2-cyclopenten-1-ol, 1r,3s-4-cyclopentene-1,3-diol 1-acetate, 1r,4s-cis-4-hydroxy-2-cyclopentenyl acetate, 5beta-acetoxycyclopentene-3beta-ol
10154103
[(1R,4S)-4-hydroxycyclopent-2-en-1-yl] acetate
CC(=O)OC1CC(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 43°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Specific Rotation +67° to +69° (20°C, 589nm) ((c=2.3 in chloroform))
Formula Weight 142.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material (1R, 3S)-(+)-4-Cyclopentene-1, 3-diol 1-acetate, stabilzed
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'