missing translation for 'onlineSavingsMsg'
Learn More

(1R,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee, Thermo Scientific Chemicals

Catalog Number p-70138
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
489446-85-7
C11H19NO4
229.27
MFCD02259727
RNJQBGXOSAQQDG-HTQZYQBOSA-N
1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid
1512527
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O
This item is not returnable. View return policy
489446-85-7
C11H19NO4
229.27
MFCD02259727
RNJQBGXOSAQQDG-HTQZYQBOSA-N
1r,3r-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-n-boc-1-aminocyclopentane-3-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1s,3r-+-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, 1r,3r-n-boc-3-aminocyclopentane carboxylic acid, 1r,3r-3-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1r,3r-3-tert-butoxy carbonylamino cyclopentanecarboxylic acid
1512527
(1R,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117.3°C
CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Assay Percent Range 94% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble
Formula Weight 229.27
Percent Purity 95%,98% ee
Quantity 500 mg
Physical Form Powder
Chemical Name or Material (1R, 3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'