missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,2S)-(+)-cis-1-Amino-2-indanol, 98%, ACROS Organics™
£34.26 - £210.00
Chemical Identifiers
CAS | 136030-00-7 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00216656 |
InChI Key | LOPKSXMQWBYUOI-DTWKUNHWSA-N |
Synonym | 1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol |
PubChem CID | 2725045 |
IUPAC Name | (1R,2S)-1-amino-2,3-dihydro-1H-inden-2-ol |
SMILES | C1C(C(C2=CC=CC=C21)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10793003
|
Acros Organics
297530010 |
1g | Glass bottle |
£34.26
1g |
In stock, ready to ship: 21 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10078611
|
Acros Organics
297530100 |
10g | Glass bottle |
£210.00
10g |
Estimated Shipment 01-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
136030-00-7 | |
149.19 | |
LOPKSXMQWBYUOI-DTWKUNHWSA-N | |
2725045 | |
C1C(C(C2=CC=CC=C21)N)O |
C9H11NO | |
MFCD00216656 | |
1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol | |
(1R,2S)-1-amino-2,3-dihydro-1H-inden-2-ol |
Specifications
(1R, 2S)-(+)-cis-1-Amino-2-indanol | |
Glass bottle | |
+ 44.5 | |
117°C to 121°C | |
98% | |
97.5 | |
98% | |
MFCD00216656 | |
Solubility in water: soluble | |
C1C(C(C2=CC=CC=C21)N)O | |
149.19 | |
149.19 | |
98% |
Authentic | |
+44.5° (20°C c=1,methanol) | |
White to Beige | |
1g | |
136030-00-7 | |
100.0 | |
C9H11NO | |
1r,2s-1-amino-2-indanol, 1r,2s-1-amino-2,3-dihydro-1h-inden-2-ol, cis-1-amino-2-indanol, 1r,2s-+-cis-1-amino-2-indanol, cis-1-amino-2,3-dihydro-1h-inden-2-ol, 1r,2s-+-1-amino-2-hydroxyindan, 1r,2s-1-amino-2-hydroxyindane, 1r,2s-+-1-amino-2-indanol, 1r,2s-+-cis-1-aminoindan-2-ol, 1r,2s-cis-1-amino-2-indanol | |
LOPKSXMQWBYUOI-DTWKUNHWSA-N | |
(1R,2S)-1-amino-2,3-dihydro-1H-inden-2-ol | |
2725045 | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (1R,2S)-(+)-cis-1-Amino-2-indanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.