missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulfinate, 98%, Thermo Scientific Chemicals

Catalog Number 11915751
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1517-82-4
C17H26O2S
294.46
NQICGNSARVCSGJ-DSRNLFJRSA-N
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
101348873
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C
This item is not returnable. View return policy
1517-82-4
C17H26O2S
294.46
NQICGNSARVCSGJ-DSRNLFJRSA-N
1r,2s,5r---menthyl s-p-toluenesulfinate, 4-methylbenzenesulfinic acid 1r,5r-2-isopropyl-5-methylcyclohexyl ester
101348873
[(1R,5R)-5-methyl-2-propan-2-ylcyclohexyl] 4-methylbenzenesulfinate
CC1CCC(C(C1)OS(=O)C2=CC=C(C=C2)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102.0°C to 104.0°C
Color Brown to Yellow or Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −190° to −204° (20°C, 589nm) (c=2, acetone)
Formula Weight 294.46
Percent Purity 98%
Physical Form Solution
Chemical Name or Material (1R, 2S, 5R)-(-)-Menthyl (S)-p-toluenesulfinate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'