Allsigns International™ 1mm Rigid Plastic Security Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA102/PVC.

Safety sign WA102/PVC hazard warning danger razorwire 200mm x 300mm 1mm rigid plastic Allsigns,

UNSPSC: 55121700

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12365638

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
27-02-2017
Add to basket


Description and Specification

Specification

Material 1mm Rigid Plastic
Message Danger razor wire
Quantity 1Each
Type Security Safety Sign
Dimensions (L x W) 200 x 300mm