Allsigns International™ 1mm Rigid Plastic Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: RG2/R.

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12319827

Quantity Price
1 £ 19.560000000000002 / Each
Estimated Shipment
05-05-2017
Add to basket


Description and Specification

Specification

Material Rigid Plastic
Message Refuge fire exit with running man/up arrow
Type Safety sign
Quantity 1Each
Dimensions (L x W) 400 x 150mm
For Use With (Application) Used to indicate suitable exit routes and safe areas for disabled people
Documentation

Technical documents