Allsigns International™ 1mm Rigid Plastic Safety Sign


Overview
Brand: Allsigns International™

Manufacturer Part Number: SA28/R.

UNSPSC: 55121712

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12379817

Quantity Price
1 £ 25.34 / Each
Estimated Shipment
10-04-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Type Safety sign
Material Rigid Plastic
Certifications/Compliance EEC 92/58/EEC
Message Fire exit with right running man
Dimensions (L x W) 400 x 200mm
Documentation

Technical documents